ContactUsLinkComponent
EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
Catalog #: 75878-132
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
KIT GLYCEROL ASY ENZYCHROM 2 UM 200 TEST
EnzyChrom™ Glycerol Assay Kit, BioAssay Systems
Catalog #: 75878-132
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glycerol Assay Kit
  • Size:
    200 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological, food, beverage etc.
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-132
  • Supplier No.:
    EGLY-200
Specifications
About this item
For quantitative determination of glycerol and evaluation of drug effects on glycerol metabolism.
  • Linear detection range 10 to 1000 μM (92 μg/dL to 9.2 mg/dL) glycerol for colorimetric assays and 2 to 50 μM for fluorimetric assays
  • Homogeneous high-throughput assay
Glycerol [Glycerin or Glycerine, C3H5 (OH) 3] is widely used in foods, beverages and pharmaceutical formulations
BioAssay Systems’ glycerol assay uses a single Working Reagent that combines glycerol kinase, glycerol phosphate oxidase and color reactions in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to glycerol concentration in the sample.
It is also a main by-product of biodiesel production. Simple, direct and automation-ready. Procedures for measuring glycerol concentrations find wide applications.
Customers Who Bought This Also Bought