Quartz Tubing, Technical Glass Products
Supplier: Technical Glass Products
Certificates
About this item
Fused Quartz consists of high-purity Silica (SiO₂) with a purity level of 99.998%.
- Extremely high purity
- High softening temperature (1670 °C)
- Optical transparency over a wide range, low thermal expansion, excellent thermal shock
- Resistance to a large number of elements and compounds
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic.
Tubing manufactured from this material is invaluable in research and industry applications.
Customers Who Bought This Also Bought