ContactUsLinkComponent
EnzyChrom™ Creatine Kinase Assay Kit, BioAssay Systems
EnzyChrom™ Creatine Kinase Assay Kit, BioAssay Systems
  75878-076
 :  
undefined
EnzyChrom™ Creatine Kinase Assay Kit, BioAssay Systems
  75878-076
 :  ECPK-100
 :  
  • Description:
    Creatine Kinase Assay Kit
  • Size:
    100 assays
  • Method:
    OD340 nm
  • Sample:
    Serum, plasma, etc.
  • Processing time:
    Five minutes
  • Cat. no.:
    75878-076
  • Supplier No.:
    ECPK-100
For quantitative determination of creatine kinase activity and evaluation of drug effects on CK activity.
  • Detection range 5 to 300 U/L creatine kinase in 96-well plate assay
  • Can be readily automated to test thousands of samples per day
Creatine kinase (CK), also known as creatine phosphokinase (CPK), is an enzyme (EC 2.7.3.2) expressed predominantly in skeletal muscle, smooth muscle and the brain
BioAssay Systems’ EnzyChrom™ Creatine Kinase Assay Kit is based on enzyme coupled reactions in which creatine phosphate and ADP is converted to creatine and ATP by CK, the generated ATP is used to phosphorylate glucose by hexokinase to generate glucose-6-phosphate, which is then oxidized by NADP in the presence of glucose-6-phosphate dehydrogenase. The produced NADPH, measured at 340 nm, is proportionate to the CK activity in the sample.
The CK enzyme consists of two subunits, which can be either B (brain type) or M (muscle type), and hence three different isoenzymes: CK-MM, CK-BB and CK-MB. CK catalyzes the conversion of creatine to phosphocreatine, consuming adenosine triphosphate (ATP) and generating adenosine diphosphate (ADP) and the reverse reaction. CK is often determined routinely in emergency patients with chest pain and acute renal failure. Elevation of CK is an indication of damage to muscle and has been associated with injury, rhabdomyolysis, myocardial infarction, myositis, myocarditis, malignant hyperthermia and neuroleptic malignant syndrome, etc. Lower levels can be an indication of alcoholic liver disease and rheumatoid arthritis. Simple, direct and automation-ready. Procedures for measuring CK activity are very desirable.