ContactUsLinkComponent
EnzyChrom™ Choline Assay Kit, BioAssay Systems
EnzyChrom™ Choline Assay Kit, BioAssay Systems
Catalog # 75878-064
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Choline Assay Kit, BioAssay Systems
Catalog # 75878-064
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  ECHO-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Choline Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, tissue, cells etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-064
  • Supplier No.:
    ECHO-100
Specifications
About this item
For quantitative determination of choline and evaluation of drug effects on choline metabolism.
  • Linear detection range colorimetric assay 1 to 100 μM, fluorimetric assay 0.2 to 10 μM choline
Choline and its metabolites play important roles in membrane structure integrity, cellular signaling and cholinergic neurotransmission
BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring choline in biological samples. In this assay, free choline is oxidized by choline oxidase to βine and H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the choline concentration in the sample.
Aberrant regulation in choline metabolism has been associated with mental illness such as anxiety.