To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Cholesterol Assay Kit
- Size:100 assays
- Method:OD340 nm
- Sample:Serum, plasma, etc.
- Processing time:40 minutes
- Cat. no.:75878-058
- Supplier No.:ECCH-100
Specifications
About this item
For quantitative colorimetric determination of cholesterol and evaluation of drug effects on cholesterol metabolism.
- Linear detection range 5 - 300 mg/dL cholesterol in 96-well plate assay
- Convenient
- Room temperature assay
- No 37°C heater is needed
- Can be readily automated to process thousands of samples per day
Cholesterol is a sterol and lipid present in the cell membranes, and is transported in the bloodstream of all animals
BioAssay Systems’ EnzyChrom™ Cholesterol Assay is based on cholesterol esterase hydrolysis of cholesterol esters to form free cholesterol and cholesterol dehydrogenase catalyzed conversion of cholesterol to cholest-4-ene-3-one, in which NAD is reduced to NADH. The optical density of the formed NADH at 340 nm is directly proportionate to the cholesterol concentration in the sample.
It is used to form cell membranes and hormones, and plays important roles in cell signaling processes. Elevated levels (hypercholesterolemia) have been associated with cardiovascular diseases such as atherosclerosis; whereas, low levels (hypocholesterolemia) may be linked to depression, cancer and cerebral hemorrhage. Simple, direct and automation-ready. Procedures for measuring cholesterol are very desirable.