ContactUsLinkComponent
EnzyChrom™ Amylase Assay Kit, BioAssay Systems
EnzyChrom™ Amylase Assay Kit, BioAssay Systems
Catalog # 75878-054
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Amylase Assay Kit, BioAssay Systems
Catalog # 75878-054
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  ECAM-100
CAS Number:  
Specifications
  • Description:
    Amylase Assay Kit
  • Size:
    100 assays
  • Method:
    OD585 nm
  • Sample:
    Blood, saliva, urine, agriculture
  • Processing time:
    40 minutes
  • Cat. no.:
    75878-054
  • Supplier No.:
    ECAM-100
Specifications
About this item
For quantitative determination of alpha-amylase activity.
  • Linear detection range 0.3 to 50 U/L α-amylase in 96-well plate assay
Amylase belongs to the family of glycoside hydrolase enzymes that break down starch into glucose molecules by acting on α-1,4-glycosidic bonds
BioAssay Systems’ EnzyChrom™ α-amylase assay method involves two steps: (1). α-amylase in the sample hydrolyzes starch and the product is rapidly converted to glucose by α-glucosidase and hydrogen peroxide by glucose oxidase; (2). hydrogen peroxide concentration is determined with a colorimetric reagent.
The α-amylases (EC 3.2.1.1) cleave at random locations on the starch chain, ultimately yielding maltotriose and maltose, glucose and "limit dextrin" from amylose and amylopectin. In mammals, α-amylase is a major digestive enzyme. Increased enzyme levels in humans are associated with salivary trauma, mumps due to inflammation of the salivary glands, pancreatitis and renal failure. Simple, direct and automation-ready. Procedures for measuring amylase activity are very desirable.