Specifications
- Description:Glucose Assay Kit
- Size:100 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Serum, plasma, urine, saliva, milk, culture medium, food, agriculture etc.
- Processing time:40 minutes
- Cat. no.:75878-052
- Supplier No.:EBGL-100
Specifications
About this item
For quantitative colorimetric/fluorometric determination of glucose and evaluation of drug effects on glucose metabolism.
- Linear detection range 5 to 300 μM (90 μg/dL to 5.4 mg/dL) glucose for colorimetric assays and 1 to 30 μM for fluorimetric assays
Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders
BioAssay Systems’ glucose assay kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to glucose concentration in the sample.
Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism. Simple, direct and high-throughput assays for measuring glucose concentrations find wide applications in research and drug discovery.