ContactUsLinkComponent
Syringe Extender, S17 Science
Syringe Extender, S17 Science
Catalog # 470102-726
Supplier:  S17 SCIENCE LLC.
undefined
Syringe Extender, S17 Science
Catalog # 470102-726
Supplier:  S17 SCIENCE LLC.
Supplier Number:  EQ 077
Specifications
  • Description:
    Syringe Extender with Luer Lock
  • Cat. no.:
    470102-726
  • Supplier No.:
    EQ 077
Specifications
About this item
The extender is used to make an air-tight connection between a syringe and plastic, vinyl, or rubber tubing.