ContactUsLinkComponent
QuantiFluo™ Ammonia/Ammonium Assay Kit, BioAssay Systems
QuantiFluo™ Ammonia/Ammonium Assay Kit, BioAssay Systems
Catalog # 75878-008
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ Ammonia/Ammonium Assay Kit, BioAssay Systems
Catalog # 75878-008
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DNH3-200
CAS Number:  
Specifications
  • Description:
    Ammonia/Ammonium Assay Kit
  • Size:
    200 assays
  • Method:
    FL360/450 nm
  • Sample:
    Biological (e.g. urine) and environmental samples.
  • Processing time:
    15 minutes
  • Cat. no.:
    75878-008
  • Supplier No.:
    DNH3-200
Specifications
About this item
For quantitative fluorometric determination of ammonia/ammonium concentration.
  • Linear detection range of 0.012 - 1 mM ammonia
  • Can be readily automated to process thousands of samples per day
Ammonia (NH3) or its ion form ammonium (NH4+) is an important source of nitrogen for living systems and is ubiquitously present in the nature
BioAssay Systems’ ammonia/ammonium assay is based on an improved o-phthalaldehyde method. This reagent reacts with ammonia/ ammonium and forms a fluorescent product. The fluorescence intensity (λex/em = 360/450nm) is proportional to the ammonia concentration in the sample.
Simple, direct and automation-ready. Procedures for measuring NH3 are very desirable.