Order Entry
Canada
ContactUsLinkComponent
Ethanol Assay Kit, BioAssay Systems
Ethanol Assay Kit, BioAssay Systems
Catalog # 75878-096
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
Ethanol Assay Kit, BioAssay Systems
Catalog # 75878-096
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DIET-500
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Ethanol Assay Kit
  • Size:
    500 assays
  • Method:
    OD580 nm (Chemical)
  • Sample:
    Alcoholic beverages etc.
  • Processing time:
    10 minutes
  • Brand name:
    QuantiChrom™
  • Cat. no.:
    75878-096
  • Supplier No.:
    DIET-500

Specifications

About this item

determination of ethanol and alcohol metabolism

  • Sensitive and accurate. Detection range 0.04 - 4% alcohol in 96-well plate assay.
  • Convenient and high-throughput. The procedure involves adding a single working reagent, incubation for 8 min, adding a Stop Reagent, and reading the optical density. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day.
  • Versatility. Assays can be executed in 96-well plate or cuvet.
  • Method: OD580nm (Chemical)
  • Samples: Alcoholic beverages etc

Alcoholic drinks are among the daily consumed beverages. Studies have shown heavy alcohol consumption may lead to various forms of liver diseases and to increased mortality rates. Quantitative determination of alcohol (ethanol, C2H5OH) finds applications in basic research, drug discovery, clinic studies and winery. Simple, direct and automation-ready procedures for measuring ethanol concentration are very desirable. BioAssay Systems’ QuantiChrom™ ethanol assay kit is based on an improved dichromate method, in which dichromate is reduced by ethanol to a bluish chromic (Cr3+) product. The intensity of color, measured at 580 nm, is a direct measure of the alcohol concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.