Order Entry
Canada
Orders LinkContactUsLinkComponent
Learning Advantage Geo Mirror

Learning Advantage Geo Mirror

Geo Mirror, 4", 6"
Catalog # 470046-686
Supplier #:  M1170
Learning Advantage Geo Mirror
Catalog # 470046-686
Learning Advantage Geo Mirror

Geo Mirror, 4", 6"

Price
$12.50
Log in for availability
Each Box of 100
Product Details & Documents

Clearly reflect images for lessons on symmetry, congruency, lines and planes.

  • Help students draw angles
  • Durable plastic

This simple yet useful teaching aide reflects shapes and images, helping students draw angles, bisections, perpendicular and parallel lines. Purple translucent polystyrene plastic is durable and student-friendly. Designed for both left and right-handed students.

Specifications
  • Description:
    Geo Mirror
  • Length:
    6"
  • Width:
    4"