Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Total Pure NGS, Omega Bio-Tek
Mag-Bind® Total Pure NGS, Omega Bio-Tek
Supplier:  Omega Bio-Tek
Certificates
undefined
Mag-Bind® Total Pure NGS, Omega Bio-Tek
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® TotalPure NGS

Specifications

Related Information

Frequently Bought Together