JS-5.9 Swinging Bucket Rotor, Beckman Coulter®
Catalog # CABK369331
Supplier: Beckman Coulter
Specifications
- Description:JS-5.9 Swinging Bucket Rotor
- Cat. no.:CABK369331
- Maximum Speed:5900 rpm
- Maximum RCF:6570×g
- Supplier No.:369331
Specifications
About this item
High-throughput processing (DNA or RNA kits), and serial dilution of small volumes.