QuantiChrom™ ATPase Assay Kit, BioAssay Systems
Catalog # 75877-962
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:ATPase Assay Kit
- Size:200 assays
- Method:OD620 nm
- Sample:ATPase and GTPase
- Processing time:30 minutes
- Cat. no.:75877-962
- Supplier No.:DATG-200
Specifications
About this item
For quantitative determination of ATPase or GTPase activity and high-throughput screen for their inhibitors.
- High sensitivity. Detects 0.007 U/L ATPase or GTPase activity
- Homogeneous "mix-and-measure" assay
- Can be readily automated for running tens of thousands of samples per day
ATPases and GTPases catalyze the decomposition of ATP or GTP into ADP or GDP and free phosphate ion
BioAssay Systems’ QuantiChrom™ ATPase/GTPase Assay Kit offers a highly sensitive method for determining ATPase/GTPase activities in a microplate format.Its proprietary formulation features a single reagent for accurate determination of enzyme activity in 30 min at room temperature. The improved malachite green reagent forms a stable dark green color with liberated phosphate, which is measured on a plate reader (600 - 660 nm).
These enzymes play key roles in transport, signal transduction, protein biosynthesis and cell differentiation.