QuantiChrom™ Mannosidase Assay Kit, BioAssay Systems
Catalog # 75877-956
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:Mannosidase Assay Kit
- Size:100 assays
- Method:OD405 nm
- Sample:Biological samples e.g. plasma, serum, tissue and culture media.
- Processing time:10 minutes
- Cat. no.:75877-956
- Supplier No.:DAMA-100
Specifications
About this item
For quantitative determination of alpha-mannosidase enzyme activity.
- Linear detection range 1 to 250 U/L for a 10-minute reaction
- This homogeneous "mix-incubate-measure" type assay can be readily automated for processing thousands of samples per day
α-Mannosidase (AMA) is an enzyme which catalyzes the cleavage of the α form of mannose. α-Mannosidase assists in the breakdown of complex sugars from glycoproteins in the lysosome. Defective AMA or deficient AMA activity causes α-mannosidosis and leads to deterioration of the central nervous system in children. BioAssay Systems’ non-radioactive, colorimetric AMA assay is based on the cleavage of 4-nitrophenol from the synthetic substrate. Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at 405 nm after addition of the stop reagent is directly proportional to the enzyme activity.