Order Entry
Canada
ContactUsLinkComponent
QuantiChrom™ Alkaline Phosphatase Assay Kit, BioAssay Systems
QuantiChrom™ Alkaline Phosphatase Assay Kit, BioAssay Systems
Catalog # 75877-954
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Alkaline Phosphatase Assay Kit, BioAssay Systems
Catalog # 75877-954
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DALP-250
CAS Number:  

Specifications

  • Description:
    Alkaline Phosphatase Assay Kit
  • Size:
    250 assays
  • Method:
    OD405 nm
  • Sample:
    Serum, plasma, etc.
  • Processing time:
    Four minutes
  • Cat. no.:
    75877-954
  • Supplier No.:
    DALP-250

Specifications

About this item

For quantitative colorimetric determination of alkaline phosphatase (ALP) activity.

  • Use only 5 μL serum or plasma sample. Linear detection range 2 - 800 U/L
  • Simple "mix-and-measure"
  • Procedure allows reliable quantitation of ALP activity within five minutes
  • All reagents are compatible with high-throughput liquid handling instruments

Alkaline phosphatase (ALP) catalyzes the hydrolysis of phosphate esters in an alkaline environment, resulting in the formation of an organic radical and inorganic phosphate

BioAssay Systems’ QuantiChrom™ Alkaline Phosphatase Assay Kit is designed to measure ALP activity directly in biological samples without pretreatment. The improved method utilizes p-nitrophenyl phosphate that is hydrolyzed by ALP into a yellow colored product (maximal absorbance at 405nm). The rate of the reaction is directly proportional to the enzyme activity.

In mammals, this enzyme is found mainly in the liver and bones. Marked increase in serum ALP levels, a disease known as hyperalkalinephosphatasemia, has been associated with malignant biliary obstruction, primary biliary cirrhosis, primary sclerosing cholangitis, hepatic lymphoma and sarcoidosis. Simple, direct and automation-ready. Procedures for measuring ALP activity in serum are becoming popular in Research and Drug Discovery.