Specifications
- Description:Nitric Oxide Assay Kit
- Size:100 assays
- Method:OD540nm
- Sample:Plasma, serum, urine, tissue/cells and foods.
- Processing time:40 minutes
- Cat. no.:75877-946
- Supplier No.:D2NO-100
Specifications
About this item
For quantitative determination of nitric oxide and evaluation of drug effects on its metabolism.
- Detection range 0.6 - 200 μM in 96-well plate
- Rapid and reliable
- Can be readily automated to measure thousands of samples per day
Nitric oxide (NO) is a reactive radical that plays an important role in many key physiological functions
QuantiChrom™ Nitric Oxide Assay Kit is designed to accurately measure NO production following reduction of nitrate to nitrite using improved Griess method. The procedure is simple and the time required for sample pretreatment and assay is reduced to as short as 30 min.
NO, an oxidation product of arginine by nitric oxide synthase, is involved in host defense and development, activation of regulatory proteins and direct covalent interaction with functional biomolecules. Simple, direct and automation-ready. Procedures for measuring NO are becoming popular in Research and Drug Discovery. Since NO is oxidized to nitrite and nitrate, it is common practice to quantitate total NO2-/NO3- as a measure for NO level. BioAssay Systems.