Spectrophotometer Cuvettes, Chemglass
Supplier: Chemglass
Certificates
About this item
Disposable cuvettes, made of polystyrene (PS) or polymethyl methacrylate (PMMA), are an excellent choice for most spectrophotometry applications in the visible range.
- Provide accurate, reliable results, even at wavelengths as low as 300 nm
- Macro and semi-micro sizes to accommodate most sample volumes
- Include an arrow mark to indicate direction of transmission and reduce variation
- Compatible with most spectrophotometers and photometers
Customers Who Bought This Also Bought