Screw Caps, Open Top, Chemglass
Supplier: Chemglass
Certificates
Certificates
About this item
Open top polypropylene screw caps supplied with PTFE-faced silicone septa.
Packaging: Supplied in packages of 144.
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
649,44 $ to 2 496,86 $
Glucose-Glutamic Acid Solution, Standard Check Solution for Biochemical Oxygen Demand (BOD), Ricca Chemical Company
Supplier: Ricca Chemical
202,41 $
tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granular BP, Ph. Eur., JP, USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
3 278,39 $
Camco® Erado-Sol® Biological and Chemical Stain Remover, Cambridge
Supplier: Cambridge Diagnostic Products
171,89 $
Lacey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
185,40 $ to 220,62 $