Mops Buffer, 10X, Teknova
Supplier: Teknova
About this item
10X MOPS Buffer for EZ Rich defined medium kit
Repeated freeze thaw causes precipitation and a reduction in cell growth. Aliquot contents upon intial thaw
Suitable for molecular biology, protein chemistry, and biochemical applications.
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
10X MOPS Buffer for EZ Rich defined medium kit
Repeated freeze thaw causes precipitation and a reduction in cell growth. Aliquot contents upon intial thaw
Suitable for molecular biology, protein chemistry, and biochemical applications.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA77443-134
- CA200059-678
- Storage temperature:
- Frozen
- Frozen
- Shipping temperature:
- Frozen
- Frozen
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- CA77443-13410X Mops buffer, sterile solutionSupplier Number: M2101Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New Product
More
Specifications
Cat. No.CA77443-134Storage temperatureFrozenShipping temperatureFrozenNew Product - CA200059-67810X Mops buffer, sterile solution10X Mops buffer, sterile solution100 mlDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.CA200059-678Storage temperatureFrozenShipping temperatureFrozen