About this item
The 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.
Type: Primary
Antigen: CD16 / CD32
Clonality: Monoclonal
Clone: 2.4G2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75875-690
- 75875-872
- 75875-716
- Antigen name:
- CD16 and CD32 molecule
- CD16 and CD32 molecule
- CD16 and CD32 molecule
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 2.4G2
- 2.4G2
- 2.4G2
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 14130/14131
- 14130/14131
- 14130/14131
- Isotype:
- IgG2b
- IgG2b
- IgG2b