Order Entry
Canada
ContactUsLinkComponent
LaboPlast® Disposable Scraper, Bürkle
LaboPlast® Disposable Scraper, Bürkle
Catalog # 75876-070
Supplier:  BURKLE INC
undefined
LaboPlast® Disposable Scraper, Bürkle
Catalog # 75876-070
Supplier:  BURKLE INC
Supplier Number:  5378-0030

Specifications

  • Description:
    LaboPlast® Disposable Scraper
  • Dimensions:
    200L x 80W mm (3.15 x 7.87")
  • Cat. no.:
    75876-070
  • Supplier No.:
    5378-0030

Specifications

About this item

The LaboPlast® Disposable Scraper, made of polystyrene, is produced in a class 7 cleanroom (10,000) and individually packaged for disposable use.

  • Polystyrene construction
  • Individually packaged

Sturdy and convenient scraper for disposable use. The scraper is suitable for scraping off, smoothing, or cleaning, as well as removing residue.

Packaging: 10 scrapers per package.