You searched for: Sera (Serum)
Sera, whether bovine, human or from other sources, provides mammalian cells with the vital growth factors, lipids and more required for their growth in culture. The type of sera required is determined by both the cells being used and the media those cells require. Serum is a traditional supplement used in cell culture. Serum provides critical nutrients and growth factors needed by the cells for optimal growth. Fetal Bovine Serum (FBS) is the most common serum used for this purpose; however, there are other serum types used in the culture of cells. Serum alternatives are also available to support cell growth if applicable for the cell type. VWR offers a wide variety of sera to meet the needs of cell culturists. Please select from the attributes below to find the sera options best suited to your application needs.
Avantor® Seradigm, Ultra Low IgG Fetal Bovine Serum (FBS)
Supplier: VWR
Ultra Low IgG Fetal Bovine Serum (FBS) is purified of immunoglobulin (IgG) using a chromatographic process that delivers significantly reduced levels of IgG (≤5μg/mL).
Expand 4 Items
Avantor® Seradigm, Premium Grade Fetal Bovine Serum (FBS)
Supplier: VWR
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications.
Expand 6 Items
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Supplier: VWR
Avantor Seradigm Select Grade FBS provides superior performance and consistency in cost-sensitive applications.
Expand 6 Items
Avantor® Seradigm, Ultimate Grade Fetal Bovine Serum (FBS)
Supplier: VWR
Ultimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
Expand 6 Items
Goat Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal goat serum is used as a blocking agent or as a control for immunoassays
Expand 1 Items
Llama Serum, Immunoreagents
Supplier: Immunoreagents
ImmunoReagents’ normal llama serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Bovine Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal bovine serum is used as a blocking agent or as a control for immunoassays
Expand 1 Items
Rabbit Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal rabbit serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Human Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal human serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Sheep Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal sheep serum is used as a blocking agent or as a control for immunoassays
Expand 1 Items
HORSE SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Heat Inactivated Sterile Serum
Expand 3 Items
HAMSTER SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Heat Inactivated Sterile Serum
Expand 3 Items
MOUSE SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Heat Inactivated Sterile Serum
Expand 3 Items
Guinea Pig Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal guinea pig serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Chicken Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal chicken serum is used as a blocking agent or as a control for immunoassays
Expand 1 Items
Canine Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal canine serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Pig Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal pig serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Horse Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal horse serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
Rat Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal rat serum is used as a blocking agent or as a control for immunoassays
Expand 1 Items
Mouse Serum, ImmunoReagents
Supplier: Immunoreagents
ImmunoReagents’ normal mouse serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
RAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Heat Inactivated Sterile Serum
Expand 3 Items
Syrian Hamster Serum, Immunoreagents
Supplier: Immunoreagents
ImmunoReagents’ normal syrian hamster serum is used as a blocking agent or as a control for immunoassays
Expand 2 Items
GOAT SERUM (sterile) (Heat Inactivated), Rockland Immunochemicals
Supplier: Rockland Immunochemical
Heat Inactivated Sterile Serum
Expand 2 Items
Rabbit Complement, MP Biomedicals
Supplier: MP Biomedicals
Product is rabbit serum, rapidly processed to preserve complement activity. Pooled rabbit serum is filtered through a 0.22 µm
Expand 1 Items
Corning® Bovine calf serum, 1 L, iron-fortified, United States origin, Corning
Supplier: Corning
Bovine calf serum, United States origin are collected at government approved abattoirs in the United States.
Expand 1 Items
Sterile Human Serum, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Sterile Serum
Expand 1 Items
Fetalgro Bovine Growth Serum, RMBIO
Supplier: Rocky Mountain Biologicals
Fetalgro® is Rocky Mountain Biologicals proprietary bovine growth serum (BGS) supplemented with additives that match or outperform FBS' cell growth capabilities in many cell lines.
Expand 1 Items
Guinea Pig Complement, MP Biomedicals
Supplier: MP Biomedicals
Product is guinea pig serum, rapidly processed to preserve complement activity.
Expand 2 Items
Corning® Donor Horse Serum, US Sourced, Corning
Supplier: Corning
Horse serum is often used in testing media for mycoplasma contamination or as a blocking agent in immunoassays
Expand 1 Items
Corning® Tetracycline Negative FBS, US Sourced
Supplier: Corning
Fetal Bovine Serum often contains the antibiotic tetracycline (tet) and researchers using very sensitive tet-inducible gene expression systems encounter problems with unintended expression as a result.