Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Multi-Element Standard, 4 Components, RICCA Chemical Company
VeriSpec® Multi-Element Standard, 4 Components, RICCA Chemical Company
Catalog # RCRV010675100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Multi-Element Standard, 4 Components, RICCA Chemical Company
Catalog # RCRV010675100N
Supplier:  Ricca Chemical
Supplier Number:  RV010675-100N
CAS Number:  

Specifications

  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010675100N
  • Supplier No.:
    RV010675-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

100 ppm: Mo, Sb, Sn, Ti

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility