Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Phosphorus Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
VeriSpec® Phosphorus Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
Catalog # RCRV010608100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Phosphorus Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
Catalog # RCRV010608100N
Supplier:  Ricca Chemical
Supplier Number:  RV010608-100N
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010608100N
  • Element:
    Phosphorus
  • Supplier No.:
    RV010608-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility