Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Cesium Standard for ICP/MS 10 ppm in 2% HNO₃
VeriSpec® Cesium Standard for ICP/MS 10 ppm in 2% HNO₃
Catalogue # RCRV010583100N
Fournisseur:  Ricca Chemical
Numéro CAS:  
undefined
VeriSpec® Cesium Standard for ICP/MS 10 ppm in 2% HNO₃
Catalogue # RCRV010583100N
Fournisseur:  Ricca Chemical
Supplier Number:  RV010583-100N
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010583100N
  • Element:
    Cesium
  • Supplier No.:
    RV010583-100N

Spécifications

À propos de cet article

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility