Order Entry
Canada
ContactUsLinkComponent
 
VeriSpec® Bismuth Standard for ICP/MS 10 ppm in 2% HNO₃, Ricca Chemical Company
  RCRV010570100N
 :  
undefined
VeriSpec® Bismuth Standard for ICP/MS 10 ppm in 2% HNO₃, Ricca Chemical Company
  RCRV010570100N
 :  RV010570-100N
 :  

 

  • Concentration:
    10 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010570100N
  • Element:
    Bismuth
  • Supplier No.:
    RV010570-100N

 

 

Certified reference material (CRM) for ICP-MS

 : Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility