VeriSpec® Thulium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
RCRV010562100N
:
- Concentration:100 µg/ml
- Matrix:2% HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Cat. no.:RCRV010562100N
- Element:Thulium
- Supplier No.:RV010562-100N
Certified reference material (CRM) for ICP-MS
: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility