Order Entry
Canada
ContactUsLinkComponent
VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, RICCA Chemical Company
VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, RICCA Chemical Company
Catalog # RCRV010528100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, RICCA Chemical Company
Catalog # RCRV010528100N
Supplier:  Ricca Chemical
Supplier Number:  RV010528-100N
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HCl
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010528100N
  • Element:
    Iridium
  • Supplier No.:
    RV010528-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility