Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Indium Standard for ICP/MS 100 ppm in 2% HNO₃
VeriSpec® Indium Standard for ICP/MS 100 ppm in 2% HNO₃
Catalog # RCRV010527100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Indium Standard for ICP/MS 100 ppm in 2% HNO₃
Catalog # RCRV010527100N
Supplier:  Ricca Chemical
Supplier Number:  RV010527-100N
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010527100N
  • Element:
    Indium
  • Supplier No.:
    RV010527-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility