Order Entry
Canada
ContactUsLinkComponent
VeriSpec® Cesium Standard for ICP/MS 100 ppm in 2% HNO₃
VeriSpec® Cesium Standard for ICP/MS 100 ppm in 2% HNO₃
Catalog # RCRV010515100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® Cesium Standard for ICP/MS 100 ppm in 2% HNO₃
Catalog # RCRV010515100N
Supplier:  Ricca Chemical
Supplier Number:  RV010515-100N
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    RCRV010515100N
  • Element:
    Cesium
  • Supplier No.:
    RV010515-100N

Specifications

About this item

Certified reference material (CRM) for ICP-MS

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility