You searched for: Lubricants
Containing additives to enhance overall performance, lubricants guarantee that equipment continues to function correctly with a strong longevity. These aerosols, pastes, or liquid substances are compatible with different moving surfaces to reduce the friction and consequently, the generated heat. Keeping machine parts separate, the oil based products prevent corrosion contamination from entering the system. Lubricants are essential aspects to machine maintenance for operations to stay highly efficient without the additional replacement part costs.
VWR® Vacuum Pump Oil No.19
Supplier: VWR International
This is a superior oil based upon special crude oil stocks noted for their durability and fine lubricating qualities.
Expand 4 Items
VWR® Vacuum Pump Oil No.20
Supplier: VWR International
VWR® No. 20 is a high-quality mechanical pump fluid distilled from a straight petroleum hydrocarbon mineral oil to yield a pure high-viscosity fluid for all mechanical pump types.
Expand 2 Items
VWR® Mechanical Pump Oil No. 21
Supplier: VWR International
Designed for use in applications where reactive or corrosive gases are present.
Expand 1 Items
Stir-Lube, Ace Glass
Supplier: Ace Glass
A superior, low melting, silicone-based lubricant which liquifies at body temperature
Expand 2 Items
Hi-Lube Heavy-Duty Liquid Stirrer Lubricant, Ace Glass Incorporated
Supplier: Ace Glass
Chlorofluorocarbon grease is designed for use at speeds up to 6000rpm with Ace Glass standard glass assemblies.
Expand 1 Items
SP Wilmad-LabGlass Hi-Vac Stopcock Grease, SP Industries
Supplier: SP Industries
Stiff, non-melting silicone lubricant that maintains consistency from -40 to 260°C.
Expand 1 Items
Hydrocarbon Soluble Siliconizing Fluid, Thermo Scientific
Supplier: Thermo Scientific
Ideal for siliconizing glass surfaces.
Expand 2 Items
Pierce™ Water Soluble Siliconizing Fluid, National Scientific
Supplier: Thermo Scientific
Ideal for siliconizing glass surfaces.
Expand 1 Items
High Vacuum Grease, Ace Glass Incorporated
Supplier: Ace Glass
Dow Corning High Vacuum Grease
Expand 1 Items
Krytox GPL, Ace Glass Incorporated
Supplier: Ace Glass
Superior performance, non-contaminating, nonflammable, general purpose grease
Expand 1 Items
Krytox LVP High Vacuum Grease, Ace Glass Incorporated
Supplier: Ace Glass
This grease has very low a vapor pressure, highly inert, and nonflammable
Expand 1 Items
Hyvac Light Pump Oil, Ace Glass
Supplier: Ace Glass
Hyvac Light Pump Oil incorporates two essential high vacuum characteristics: low vapor pressure which does not materially increase at temperatures up to 50 °C, and viscosity sufficiently low for use at 15 °C
Expand 3 Items
Mineral Oil Dropper Bottle
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Used as an overlay on DNA samples to minimize evaporation.
Expand 1 Items
Vacuum Fluids for Rotary Vane Pumps, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Agilent pump fluids offer the best performance for your DS-and MS-series pumps.
Expand 1 Items
HSG-75, Electrically conductive heat sink grease, Ag ≈75%, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Thermally and electrically conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent to reduce thermal and electrical contact resistance
Expand 1 Items
HSG, Heat Sink Grease, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Thermally conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent, to reduce thermal contact resistance
Expand 2 Items
Precision® Vacuum Pump Oil, B+, Thermo Scientific
Supplier: Thermo Fisher Scientific
Double vacuum distilled to remove light fractions
Expand 1 Items
VACUUBRAND® Vacuum Pump Oil B, BrandTech
Supplier: Brandtech
Ensure the reliability of your VACUUBRAND® Rotary Vane or Chemistry-HYBRID vacuum pump by using Pump Oil B
Expand 2 Items
Hyaline Synthetic Low Temperature Cryostat Lubrication Kit, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
This product is a quality alternative to Leica and is made from the highest quality, low-temperature synthetic lubricants.
Expand 1 Items
Stopcock/Vacuum Grease
Supplier: Liveo
Highly Resistant to Water, Alkalis, and Chemicals
Expand 1 Items
Rough Pump Oil #19 for MSD Pumps, Oil Vacuum Pump, Restek
Supplier: Restek
Rough pump oil is formulated from crude oil stocks known for their durability and line-lubricating qualities.
Expand 1 Items
Injector Lubricant for Agilent HPLC Systems, Restek
Supplier: Restek
For use with Agilent 1050 and 1100 HPLC systems.
Expand 1 Items
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
A silicone lubricant for glass-to-glass joints and glass-to-rubber connections. Resistant to most chemicals and is inert and heat stable.
Expand 1 Items
PTFE-like Spray, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TFE release agent, dry lubricant. No discernible transfer, no migration, no build-up. Ideal for cold embedding.
Expand 1 Items
Synthetic Cryostat Oil, 2 oz. Bottle
Supplier: Mortech Manufacturing
Ultra low temperature synthetic cryostat oil.
Expand 1 Items
Inland 45 Pump Oil, Restek
Supplier: Restek
Recommended for optimum mass spec performance, this oil pump is compatible with buna-N, neoprene, and Viton® seals.
Expand 1 Items
Oil for Belt Driven Pumps, HyVac
Supplier: Hyvac
Ideal for belt driven vacuum pumps
Expand 1 Items
ROTAVISC Oils for Rotational Viscometers, IKA
Supplier: IKA Works
These certified standard silicone oils can be used to check the measurement accuracy of ROTAVISC.