VeriSpec™ Antimony (Sb) Standard for AAS 20 ppb in 1% HNO₃
Catalog # RCRV010127-50N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Concentration:20 ppb
- Matrix:1% HNO₃
- Pack type:Plastic Bottle
- Size:50 ml
- Cat. no.:RCRV010127-50N
- Element:Antimony
- Supplier No.:RV010127-50N
Specifications
About this item
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility