You searched for: Hot Plates
Less hazardous than baths, the hotplates uniformly heat substances for any required procedure. White ceramic surface options permit the slow titrant addition for concentration determinations and have the resistance to work with even corrosive chemicals. With superior durability to withstand accidental impacts, aluminum surface heating plates also spread high temperatures evenly. The economical hotplates are equipped with innovative safety features for efficient operation and workplace safety.
Dylatherm™ Hotplate
Supplier: VWR International
The economical and durable Dylatherm hot plates are ideal for general heating.
Expand 1 Items
VWR® Dylatherm™ Hot Plate
Supplier: VWR International
The economical Dylatherm™ hot plate features a large 16.5 cm (61/2”) diameter cast aluminum top plate that provides an even heating surface capable of heating 1500 mL of water.
Expand 1 Items
VWR® Dylatherm™ Hotplate 230 V
Supplier: VWR International
The economical Dylatherm™ Hot Plate features a large 16.5 cm (6.5") diameter cast aluminum top plate that provides an even heating surface capable of heating 1500 mL of water.
Expand 1 Items
VWR® Advanced Hotplates
Supplier: VWR International
VWR® Advanced Hotplates
Expand 2 Items
VWR® Hotplates
Supplier: VWR International
VWR® hotplates with temperature control up to 400 °C.
Expand 1 Items
VWR® Mini Hotplate 120 V
Supplier: VWR International
Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 mL of water.
Expand 2 Items
VWR® Mini Hotplates
Supplier: VWR International
These rugged, compact and reliable units are ideal for educational laboratories, and can heat up to 1 L of water.
Expand 1 Items
VWR®, Advanced Hotplates
Supplier: VWR International
Advanced hotplates are designed to deliver accurate and repeatable results in all research, academic, and industrial applications. With temperature ranges up to 500 °C the VWR® hotplates provide superior temperature control.
Expand 2 Items
Corning® Low-Profile Hot Plates, Corning
Supplier: Corning
Low-profile hot plates with a ceramic glass plate provide precision-controlled heating for laboratory applications. Microprocessor-controlled heating ensures consistent performance.
Expand 2 Items
Corning® Scholar PC-170 Hot Plates, Corning
Supplier: Corning
Economical hot plate with white enamel steel-top plate
Expand 2 Items
Corning® PC-200 Low Profile Hot Plate
Supplier: Corning
Low-profile hot plates with a ceramic glass plate provide precision-controlled heating for laboratory applications. Microprocessor-controlled heating ensures consistent performance.
Expand 1 Items
Corning® Digital Hot Plates, Corning
Supplier: Corning
Corning® hotplates with Pyroceram® glass-ceramic top, rugged housing and heating elements and digital temperature display.
Expand 10 Items
SP Bel-Art ProCulture Warming Plate 37 °C, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1 °C) outside of the incubator.
Expand 2 Items
Barnstead/Thermolyne Hot Plate, Thermo Scientific
Supplier: Thermo Fisher Scientific
For lab, clinic, classroom, or shop
Expand 2 Items
Hot Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
These hot plates feature demand-type thermostat controls that will automatically reach and hold temperature to within ±3 °C (5 °F) at 100 °C (212 °F).
Expand 3 Items
Guardian™ 2000 Analog Hot Plates and Stirrers
Supplier: Ohaus
Rugged and reliable analog hotplates and stirrers for general purpose heating and stirring.
Expand 7 Items
Thermolyne® Hotplate
Supplier: Thermo Fisher Scientific
Perfect for Small Laboratories.
Expand 1 Items
Barnstead/Thermolyne Electric Hot Plate, Type 2300, Thermo Scientific
Supplier: Thermo Fisher Scientific
For laboratories where an inexpensive heat source is needed and bench space is scarce.
Expand 1 Items
RT2 Digital Hotplate, Thermo Scientific
Supplier: Thermo Fisher Scientific
Hotplates RT2 Basic Model; 140mm top; Ceramic Coated aluminum 120V, 60Hz, 3.94x11.42x6.34in, In addition to a hot top warning indicator, an overheat prevention circuit turns off the heater if the top plate temperature reaches 450degC.
Expand 2 Items
Cimarec™ Basic Hotplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
A sleek, rugged design and construction reduces the chance of top plate breakage and promotes durability, for economical high performance.
Expand 2 Items
Remote Control Hot Plates, Type RC2200, Thermo Scientific
Supplier: Thermo Fisher Scientific
Designed for remote control operation in fume hoods or other corrosive environments.
Expand 2 Items
Stuart® Hotplates, Undergrad Series, Cole Parmer
Supplier: Argos Technologies
Get accurate surface temperature in real time.
Expand 2 Items
Digital Hotplates, SuperNuova+™, Barnstead, Thermo Scientific
Supplier: Thermo Fisher Scientific
These SuperNuova+™ hotplate series provides reliable performance, dependable safety and simple operation.
Expand 3 Items
Cimarec+™ Hotplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
The Thermo Scientific Cimarec+™ series digital hotplates are designed to provide exceptional safety, and temperature performance for routine protocols
Expand 4 Items
Guardian 5000 Hotplates, OHAUS
Supplier: Ohaus
Reliable hotplates with exceptional safety features for everyday use. Guardian 5000 hotplates are designed for safety and simplicity in all research, academic and industrial applications.
Expand 2 Items
Portable Gas Hot Plate, Global Market Connections
Supplier: Global Market Connections
This gas operated hot plate provides a fully adjustable infrared heat source of up to 1100°C (2000°F) without the need for an electrical outlet
Expand 1 Items
Corning® Scholar PC-170 Hot Plate
Supplier: Corning
Most Economical Hot Plate.
Expand 1 Items
Histo Orientator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
The convenient 51×51 mm (2×2") heated surface can be used for either 'de-wrinkling' of paraffin sections or for the 'rapid attachment' of fresh frozen sections. A quick touch of the slide to the hot plate will remove wrinkles and bubbles on sections. Also keep a unit on your cryostat to ensure solid attachment of frozen sections to the slide which prevents loosening of the tissue during agitation in hand staining.
Expand 1 Items
RC Series Hot Plates, Velp Scientifica
Supplier: VELP SCIENTIFIC INC.
Single-position and two-position heating plates with aluminum top and temperature regulation.
Expand 1 Items
Butane Burner Hot Plate
Supplier: G.S.C. INTERNATIONAL, INC.
Unique, compact, and affordable staple for your lab!