Semi-Micro/Macro Cuvette, PerkinElmer
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Certificates
About this item
Disposable cells are available in polystyrene and polymethyl methacrylate formats
These cells greatly reduce the risk of contamination, remove the need for cleaning reagents, and lower laboratory costs when compared to quartz or glass cells.
Also avialable either as macro or semi-micro cells.
Frequently Bought Together