Orders LinkContactUsLinkComponent
STEM Explorations: Swing Ride
STEM Explorations: Swing Ride
STEM Explorations: Swing Ride
Catalog # 470224-148
Supplier:   K'nex Industries
Supplier #:  77077
STEM Explorations: Swing Ride
Catalog # 470224-148
Supplier:  K'nex Industries
STEM Explorations: Swing Ride
STEM Explorations: Swing Ride
STEM Explorations: Swing Ride
Price
$120.55
Log in for availability
Kit of 1
Product Details & Documents
Explore STEM concepts while building a motorized Swing Ride!
  • The Experiment Guide is aligned to national STEM standards
  • Appropriate for grade levels 5-9
  • Requires 2 "AA" batteries, not included
Using the materials included in this set, middle-school aged children will be engaged and energized as they further their knowledge and understanding of the science, technology, engineering and math concepts associated with a real-life amusement park ride. The set includes over 470 parts plus a battery-powered motor for a single child or team of 2-3 children to build a working Swing Ride, plus 2 additional models: a Ferris Wheel and a Boom Ride (each model can be built one at a time). Once built, a downloadable booklet guides students through 3 hands-on, inquiry-based experiments on each of the models. Students will learn about how mass affects speed, how to gear-up and gear down, and much more!
Specifications
  • Description:
    STEM Explorations: Swing Ride