ContactUsLinkComponent
VWR® Granite Platform with Anti-Vibration Isolation
VWR® Granite Platform with Anti-Vibration Isolation
Catalog # 10799-162
Supplier:  VWR International
undefined
VWR® Granite Platform with Anti-Vibration Isolation
Catalog # 10799-162
Supplier:  VWR International
Specifications
  • Description:
    Granite platform for weighing; 70 lbs. maximum load; 38.1W×33L cm (15×13") slab
  • Environmentally Preferable:
  • Cat. no.:
    10799-162
Specifications
About this item
The VWR® Granite Platform with Anti-Vibration Isolation was designed to provide optimal performance for high precision balances in environments where vibration may affect weighing results.
  • Features high grade silicone gel vibration isolators
  • Suitable for loads up to 70 lbs. (31.75 kg)
  • No air supply required
  • Cleanroom compatible
Easy to install and maintain, the VWR® Granite Platform with Anti-Vibration Isolation is a great addition to any laboratory.