Order Entry
Canada
Orders LinkContactUsLinkComponent
BoneClones® Ankle Sprain Model

BoneClones® Ankle Sprain Model

Ankle Sprain Model, 31.8 cm, 24 cm, 8.3 cm
Catalog # 470224-440
Supplier #:  KO-334-AJ
BoneClones® Ankle Sprain Model
Catalog # 470224-440
BoneClones® Ankle Sprain Model
BoneClones® Ankle Sprain Model

Ankle Sprain Model, 31.8 cm, 24 cm, 8.3 cm

Price
$735.00
Log in for availability
Each
Product Details & Documents

Ankle sprains and ligament ruptures are common injuries among athletes and active people.

  • Four Adjustable Ligaments
  • Stand Supplied
  • Lesson Plan Supplied

In addition to being painful, they can limit an affected person's occupational and recreational activity. Therefore, it is important for clinicians to be able to conduct manipulative tests during a physical exam to determine if a ligament has been ruptured. This tactile ankle ligament rupture model will help health and medical students feel the differences between an intact ligament and a ruptured ligament. Each of the commonly ruptured ligaments of the ankle (anterior talofibular, calcaneofibular, posterior talofibular, and deltoid) can be toggled to feel the difference between an intact and a ruptured ligament.

This model was created in conjunction between BoneClones and Dr. Peter Ward of the West Virginia School of Osteopathic Medicine. A corresponding lesson plan, written by Dr. Ward, accompanies this model.

Specifications
  • Description:
    Ankle Sprain Model
  • Height:
    8.3 cm
  • Length:
    24 cm
  • Width:
    31.8 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.