Order Entry
Canada
Orders LinkContactUsLinkComponent
BoneClones® Sectioned Brain Model

BoneClones® Sectioned Brain Model

Sectioned Brain, 7, 15.9 cm, 10.8 cm
Catalog # 470224-438
Supplier #:  KO-515
BoneClones® Sectioned Brain Model
Catalog # 470224-438
BoneClones® Sectioned Brain Model
BoneClones® Sectioned Brain Model

Sectioned Brain, 7, 15.9 cm, 10.8 cm

Price
$1,610.00
Log in for availability
Each
Product Details & Documents

Sectioned along the Coronal axis, this model offers an innovative way to study the structure of the brain.

  • Coronal sections
  • Extensive lesson plan

Current diagnostic tests available allow for various views of structures in the same patient. Given that neurologic anatomical markers are often used to confirm suspected diagnoses, students in health sciences must become adept at deciphering neurologic structures from various views. Until recently, proper models of the human brain in coronal sections have not been available. We are proud to offer the Bone Clones® Human Brain in coronal sections, which has been created in consultation with Cara Davies, Ph.D. Dr. Davies has also created an extensive lesson plan, which is provided each model.

Specifications
  • Description:
    Sectioned Brain
  • Height:
    10.8 cm
  • Width:
    15.9 cm
  • No. of parts:
    7