Order Entry
Canada
Orders LinkContactUsLinkComponent
Difco™ m Enterococcus Agar, BD Diagnostics

Difco™ m Enterococcus Agar, BD Diagnostics

Supplier:   BD
Difco™ m Enterococcus Agar, BD Diagnostics
Supplier:  BD
Certificates
Difco™ m Enterococcus Agar, BD Diagnostics

1 Options Available Below

Product Details & Documents

Selective isolation and enumeration of enterococci using the membrane filtration technique.

Documents

Product Family Options

Product Information

  • Size
    Availability
    Price
  • CA90003-930
    m Enterococcus Agar
    m Enterococcus Agar
    100 g
    Discontinued
    Supplier Number: -
  • CA90003-932
    m Enterococcus Agar
    m Enterococcus Agar
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $370.00
    $370.00
    Each (500g)
       $370.00

    Supplier Number: 274620