Difco™ KF Streptococcus Agar, BD Diagnostics
CA90003-436
: BD
:
- Description:KF Streptococcus Agar
- Size:500 g
- Cat. no.:CA90003-436
- Supplier No.:249610
Use with TTC or TTC Solution 1%, if desired For selective enumeration of fecal streptococci.