Heating Mantles, Series M, Glas-Col®
Supplier: Glas-Col
Certificates
About this item
Compact aluminum housed heating mantle.
- Fabric interior
- Durable aluminum outer shell
- Space saving compact design
Our Series "M" mantle is the most compact mantle with an aluminum shell. The resilient glass fabric interior supports the flask and protects it from thermal strain. Only the bottom of flask is covered so the contents are fully visible during process.
Delivery: 7-10 days after order.
Specifications
- Catalog No:
- 33787-622
- 33787-620
- Electrical:
- 115V
- 115V