Heating Mantles, Griffin Beakers, Glas-Col®
Supplier: Glas-Col
Certificates
About this item
Excellent fit and heating for griffin beakers.
- CSA certified
- Pliable interior fabric
- Safe small volume heating
- For use with griffin beakers
This heating mantle covers both the bottom and the sides of the beaker for even heating of all surfaces. Withstands 400 degree C internal operating temperature.
Delivery: 7-10 days after order.
Specifications
- Catalog No:
- 33795-106
- 89259-000
- 89260-026
- 33795-220
- 33795-183
- 33795-161
- 33795-128
- Electrical:
- 115V
- 115V
- 115V
- 115V
- 115V
- 115V
- 115V