Stir Control, Glas-Col®, LLC
Catalog # 33797-525
Supplier: Glas-Col
Small Business Enterprise
Specifications
- Capacity:Operates One Mantle
- Description:Stir Control, 120 V
- Cat. no.:33797-525
- For Use With:TEM Stir Mantles
- Supplier No.:104AEMC4
Specifications
About this item
Creates and synchronizes the magnetic stirring field.
- Magnetic stir control
- Synchrostart maintains link between field and bar
- LED relative field speed indication
- Fuse protection
The Stir Control II creates and synchronizes the magnetic field using our TEM stir mantle. When restarting Glas-Col's exlusive Synchrostart feature maintains the link between the field and stir bar. Control connects to Stir Mantle by cord so that it can be placed outside corrosive hood atmospheres.
Delivery: 7-10 days after order.
Packaging: Shipping weight: 2 lbs.