E.Z.N.A.® Soil DNA Kit, Omega Bio-tek
The E.Z.N.A.® Soil DNA Kit is formulated to isolate high purity cellular DNA from soil samples typically containing humic acid and other inhibitors of PCR
- Novel and proprietary method to isolate genomic DNA
- Successfully used to isolate DNA from Gram-positive and negative bacteria
- Isolated DNA can be used for most downstream applications
This kit has been successfully used to isolate DNA from Gram-positive and negative bacteria, fungi, yeast, and algae that inhabit a range of samples including clay, sandy, peaty, chalky, or loamy soil samples. Isolated DNA can be used for most downstream applications, including PCR, Southern blot and NGS analysis.
This kit uses a novel and proprietary method to isolate genomic DNA from a variety of environmental samples without organic extractions.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Soil
- Target molecule:DNA
- Sample size:200 mg–1 g
- Description:Soil DNA Isolation Kit
Frequently Bought Together