Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
Catalog #: 75838-694
Supplier: Globe Scientific
Specifications
- Description:Biohazard bag, scoreline, and document pouch
- Size:6×9"
- Cat. no.:75838-694
- Supplier No.:4921
Specifications
About this item
The special glue closure of our specimen transport bags offer a liquid-tight tamper evident seal.
- 2 mil gauge polyethylene (PE)
- Attached document compartment
- Glue closure tamper evident seal
- Imprinted with biohazard symbol
- Perfect for the transport of drug testing products
Simply pull off the liner to expose the glue adhesive and press to close.
Customers Who Bought This Also Bought