Orders LinkContactUsLinkComponent
Drip Pads, Justrite®
Drip Pads, Justrite®
Supplier:   Justrite
Drip Pads, Justrite®
Supplier:  Justrite
Certificates
Drip Pads, Justrite®
Drip Pads, Justrite®
4 Options Available Below
Product Details & Documents
Capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable - great for containing small oil drips from leaky vehicles, industrial machinery equipment, hydraulic lines, pipes, and drums.
  • Made from UV- and chemical-resistant modified PVC.
  • Patented design feature includes a coated mesh outer layer to trap coarse debris.
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs. Helps comply with EPA for spill prevention (SPCC).
Product Family Options
Product Information
  • Ext. dimensions
    Absorbency
    Weight
    Availability
    Price
  • 10861-266
    Pad Drip Standard
    Pad Drip Standard
    21 W x 23 D"
    5 L
    2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - 256,36 $
    256,36 $
    New Customer Promotional Price
    Each
       256,36 $
    Supplier Number: 28458
  • 10861-268
    Pad Drip Extended
    Pad Drip Extended
    24 W x 36 D"
    7 L
    7 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - 387,58 $
    387,58 $
    New Customer Promotional Price
    Each
       387,58 $
    Supplier Number: 28459
  • 10861-270
    Pad Drip Extended Refills
    Pad Drip Extended Refills
    24 W x 36 D"
    7 L
    7 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - 138,63 $
    138,63 $
    New Customer Promotional Price
    Each
       138,63 $
    Supplier Number: 28461
  • 10861-296
    Pad Drip Refills
    Pad Drip Refills
    21 W x 23 D"
    5 L
    2 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - 49,66 $
    49,66 $
    New Customer Promotional Price
    Each
       49,66 $
    Supplier Number: 28460