Precision Ground Stirrer Shaft, Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Glass stirrer shaft with a precision-ground surface
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Confirms to USP Type I and ASTM E438, Type I, Class A requirements.
Stirrer Bearings, PTFE
Product Information
Frequently Bought Together