Bio-Safe II Complete Counting Cocktail, RPI
Catalog # 61000-508
Supplier: RPI
Small Business Enterprise
CAS Number:
Specifications
- Description:Bio-Safe II Complete Counting Cocktail
- Size:4 L
- Cat. no.:61000-508
- Supplier No.:111195
Specifications
About this item
High performance biodegradable cocktail for counting a wide range of samples including: aqueous solutions, biological materials, salt-buffer combinations and organic solutions.
Bio-Safe II has a sample holding capacity of up to 25% aqueous volume within a clear, stable region. This large capacity can actually reduce the amount of scintillation fluid required which becomes important when using miniature vials. Bio-Safe II will not permeate plastic scintillation vials even over extended storage periods.