About this item
Ready-to-use 100 bp DNA ladder with excellent resolution of 12 different sized fragments at a great value.
- Ready-to-use DNA Ladder (pre-mixed with loading dye) - no dilution necessary
- Concentration: 100 µg / 1 ml; (Recommended loading: 0.5 µg or 5 µl)
- 12 fragments suitable for sizing double-stranded DNA from 100 bp to 3 Kb with enhanced intensity at 500 bp
- Components: 100 bp DNA Ladder (pre-mixed with loading dye) and 6X DNA Loading Dye for sample loading
- Tracking dyes: Bromophenol blue, xylene cyanol FF
- Gel Compatibility: Agarose gels
The IBI 100bp DNA Ladder, containing 12 linear double-stranded DNA fragments, is suitable for sizing double-stranded DNA from 100bp to 3,000bp
This product comes pre-mixed with loading dye and is ready to use, no dilution necessary. There is also an extra vial of 6X loading dye included in the 50µg/500µl ladder pack.
Typical load per well is 5 µl. Product is sufficient for 100 gel lanes (applications).
The 100bp DNA Ladder is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 12 fragments, suitable for use as molecular weight standards for electrophoresis. The enhanced intensity of the 500bp fragment was designed for quick and easy determination of electrophoresis results.
Specifications
- Storage conditions:25°C (6 months), 4°C (12 months), -20°C (24 months)
- Loading volume:0.5 µg (5µl)
- Shipping temperature:Approved for shipping at room temperature or dry ice